Anti-CCM2 Mouse Polyclonal Antibody
ABNOH00083605-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Cerebral Cavernous Malformation 2
- Antigen symbol:CCM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:83605
- Antigen synonyms:MGC74868|Cerebral cavernous malformations 2 protein|MGC4067|C7orf22|Malcavernin|MGC4607|Cerebral cavernous malformation 2|Ccm2|PP10187.|CCM2_HUMAN
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CCM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human