Order Entry
Slovenia
ContactUsLinkComponent
Anti-COLEC12 Mouse Monoclonal Antibody [clone: 4A7]
Anti-COLEC12 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00081035-M01
Supplier:  Abnova
undefined
Anti-COLEC12 Mouse Monoclonal Antibody [clone: 4A7]
Catalog # ABNOH00081035-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collectin sub-family member 12
  • Antigen symbol:
    COLEC12
  • Clonality:
    Monoclonal
  • Clone:
    4A7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    81035
  • Antigen synonyms:
    SCARA4|CLP1|NSR2|SRCL
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: COLEC12
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human