Anti-PCDH20 Mouse Monoclonal Antibody [clone: 2C3]
Catalog # ABNOH00064881-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protocadherin 20
- Antigen symbol:PCDH20
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:64881
- Antigen synonyms:PCDH13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PCDH20
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human