Anti-DEPTOR Rabbit Polyclonal Antibody
Catalog # ABNOH00064798-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DEP Domain Containing MTOR-interacting protein
- Antigen symbol:DEPTOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:64798
- Antigen synonyms:DEPDC6|DEP.6|DEPTOR|DEP domain-containing protein 6|DEP domain-containing mTOR-interacting protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DEPTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse