Anti-UNKL Mouse Polyclonal Antibody
ABNOH00064718-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Unkempt family zinc finger-like
- Antigen symbol:UNKL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:64718
- Antigen synonyms:ZC3HDC5L|C16orf28|ZC3H5L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: UNKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human