Anti-DCP1A Mouse Monoclonal Antibody [clone: 2D12]
Catalog # ABNOH00055802-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:decapping mRNA 1A
- Antigen symbol:DCP1A
- Clonality:Monoclonal
- Clone:2D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55802
- Antigen synonyms:Smad4-interacting transcriptional co-activator|decapping enzyme |DCP1A|Nbla00360|HSA275986|putative protein product of Nbla00360|SMIF|decapping enzyme hDcp1a|transcription factor SMIF|SMAD4IP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DCP1A
Clonality: Monoclonal
Clone: 2D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human