Anti-PRKAG2 Mouse Monoclonal Antibody [clone: 3C4]
ABNOH00051422-M01
: Abnova
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 2
- Antigen symbol:PRKAG2
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51422
- Antigen synonyms:CMH6|AAKG|AAKG2|WPWS|H91620p
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PRKAG2
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human