Anti-TBX21 Mouse Monoclonal Antibody [clone: 2F10]
ABNOH00030009-M10
: Abnova
- Antibody type:Primary
- Antigen name:T-box 21
- Antigen symbol:TBX21
- Clonality:Monoclonal
- Clone:2F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:30009
- Antigen synonyms:T-box transcription factor TBX21|TBX21 |T-cell-specific T-box transcription factor T-bet|T-PET|T-box protein 21|T-bet|TBLYM|TBET
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TBX21
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human