Anti-POLG2 Mouse Monoclonal Antibody [clone: 1A2]
Catalog # ABNOH00011232-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA) Gamma 2, Accessory Subunit
- Antigen symbol:POLG2
- Clonality:Monoclonal
- Clone:1A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11232
- Antigen synonyms:PolG-beta|p55|mitochondrial|MtPolB|Mitochondrial DNA polymerase accessory subunit|DNA polymerase gamma accessory 55 kDa subunit|POLG2|HP55|MTPOLB|DNA polymerase subunit gamma-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: POLG2
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human