Anti-AKAP11 Mouse Polyclonal Antibody
ABNOH00011215-A01
: Abnova
- Antibody type:Primary
- Antigen name:A-kinase anchor protein 11
- Antigen symbol:AKAP11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:11215
- Antigen synonyms:PPP1R44|AKAP-11|AKAP220|PRKA11
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: AKAP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human