Order Entry
Slovenia
ContactUsLinkComponent
Anti-TSHZ1 Mouse Monoclonal Antibody [clone: 2F1]
Anti-TSHZ1 Mouse Monoclonal Antibody [clone: 2F1]
  ABNOH00010194-M07
 :  Abnova
undefined
Anti-TSHZ1 Mouse Monoclonal Antibody [clone: 2F1]
  ABNOH00010194-M07
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Teashirt zinc finger homeobox 1
  • Antigen symbol:
    TSHZ1
  • Clonality:
    Monoclonal
  • Clone:
    2F1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    10194
  • Antigen synonyms:
    SDCCAG33|Teashirt homolog 1|TSH1|Antigen NY-CO-33|TSHZ1|NY-CO-33|CAA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: TSHZ1
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human