Anti-TXNDC9 Mouse Monoclonal Antibody [clone: 1E8-7C12]
Catalog # ABNOH00010190-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Thioredoxin domain containing 9
- Antigen symbol:TXNDC9
- Clonality:Monoclonal
- Clone:1E8-7C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10190
- Antigen synonyms:TXNDC9|APACD|ATP-binding protein associated with cell differentiation|PHLP3|Protein 1-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TXNDC9
Clonality: Monoclonal
Clone: 1E8-7C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human