Anti-AIM2 Mouse Polyclonal Antibody
ABNOH00009447-B01P
: Abnova
- Antibody type:Primary
- Antigen name:absent in melanoma 2
- Antigen symbol:AIM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9447
- Antigen synonyms:Ifi210,interferon-inducible protein AIM2|PYHIN4,CRYBG2,Gm1313
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: AIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human