Anti-CBFA2T2 Mouse Monoclonal Antibody [clone: 3A2]
ABNOH00009139-M05
: Abnova
- Antibody type:Primary
- Antigen name:Core-binding factor, runt Domain, alpha subunit 2; translocated to, 2
- Antigen symbol:CBFA2T2
- Clonality:Monoclonal
- Clone:3A2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9139
- Antigen synonyms:MTG8-like protein|MTG8-related protein 1|EHT|p85|CBFA2T2|ETO homologous on chromosome 20|MTGR1|Protein CBFA2T2|Myeloid translocation-related protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CBFA2T2
Clonality: Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human