Anti-PTPN4 Mouse Monoclonal Antibody [clone: 3C8]
ABNOH00005775-M01
: Abnova
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase, Non-Receptor Type 4
- Antigen symbol:PTPN4
- Clonality:Monoclonal
- Clone:3C8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5775
- Antigen synonyms:PTPase-MEG1|Megakaryocyte Phosphatase|EC 3.1.3.48
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PTPN4
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human