Order Entry
Slovenia
ContactUsLinkComponent
Anti-NDUFA2 Mouse Monoclonal Antibody [clone: 6E7]
Anti-NDUFA2 Mouse Monoclonal Antibody [clone: 6E7]
Catalog # ABNOH00004695-M01
Supplier:  Abnova
undefined
Anti-NDUFA2 Mouse Monoclonal Antibody [clone: 6E7]
Catalog # ABNOH00004695-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    NADH dehydrogenase (ubiquinone) 1, alpha subcomplex 2, 8kDa
  • Antigen symbol:
    NDUFA2
  • Clonality:
    Monoclonal
  • Clone:
    6E7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    4695
  • Antigen synonyms:
    CIB8|B8|CD14
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NDUFA2
Clonality: Monoclonal
Clone: 6E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human