Anti-ACKR1 Mouse Polyclonal Antibody
Catalog # ABNOH00002532-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Atypical Chemokine Receptor 1 (Duffy blood group)
- Antigen symbol:ACKR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2532
- Antigen synonyms:CCBP1|CD234|GpFy|DARC|GPD|WBCQ1|FY|Dfy
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human