Anti-DVL3 Mouse Monoclonal Antibody [clone: 4H2]
Catalog # ABNOH00001857-M04A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dishevelled segment polarity protein 3
- Antigen symbol:DVL3
- Clonality:Monoclonal
- Clone:4H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1857
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: DVL3
Clonality: Monoclonal
Clone: 4H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human