Anti-CACNB1 Mouse Monoclonal Antibody [clone: 1G6]
ABNOH00000782-M01
: Abnova
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, Beta 1 Subunit
- Antigen symbol:CACNB1
- Clonality:Monoclonal
- Clone:1G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:782
- Antigen synonyms:CCHLB1|CACNLB1|CAB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CACNB1
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human