Anti-ARAF Mouse Monoclonal Antibody [clone: 5H8]
ABNOH00000369-M02
: Abnova
- Antibody type:Primary
- Antigen name:A-Raf Proto-Oncogene, Serine/Threonine Kinase
- Antigen symbol:ARAF
- Clonality:Monoclonal
- Clone:5H8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:369
- Antigen synonyms:RP1-230G1.1|ARAF1|RAFA1|PKS2|A-RAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ARAF
Clonality: Monoclonal
Clone: 5H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human