Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-ALAS1 Mouse Monoclonal Antibody [clone: 3G10]
  ABNOH00000211-M01
 :  Abnova
undefined
Anti-ALAS1 Mouse Monoclonal Antibody [clone: 3G10]
  ABNOH00000211-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Delta-aminolevulinate synthase 1
  • Antigen symbol:
    ALAS1
  • Clonality:
    Monoclonal
  • Clone:
    3G10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    211
  • Antigen synonyms:
    ALAS-H|ALAS|MIG4|ALASH|ALAS3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: ALAS1
Clonality: Monoclonal
Clone: 3G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human