Anti-PIP4K2A Mouse Monoclonal Antibody [clone: OTI1B2]
ORIGTA502285
: OriGene
- Antibody type:Primary
- Antigen name:phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
- Antigen symbol:PIP4K2A
- Clonality:Monoclonal
- Clone:OTI1B2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:46 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PIP4K2A (phosphatidylinositol-5-phosphate 4-kinase, type II, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human