Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-KRT13 Goat Antibody
  ORIGTA305710
 :  OriGene
Anti-KRT13 Goat Antibody
  ORIGTA305710
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    keratin 13
  • Antigen symbol:
    KRT13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
  • Sequence:
    C-TSNASGRRTSDVRRP
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20C. Minimize freezing and thawing.
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 01-03?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: KRT13 (keratin 13)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human