Order Entry
Slovenia
ContactUsLinkComponent
Anti-KLRB1 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HP-3G10]
Anti-KLRB1 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HP-3G10]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-KLRB1 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: HP-3G10]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 65-1619-T100
  • 65-1619-T025
  • Antigen symbol:
  • KLRB1
  • KLRB1
  • Antigen name:
  • Killer cell lectin-like receptor subfamily B, member 1
  • Killer cell lectin-like receptor subfamily B, member 1
  • Antigen synonyms:
  • NKRP1A|hNKR-P1A|NKR-P1A|NKR-P1|NKR|CLEC5B|CD161
  • NKRP1A|hNKR-P1A|NKR-P1A|NKR-P1|NKR|CLEC5B|CD161
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • HP-3G10
  • HP-3G10
  • Reactivity:
  • Baboon, Rhesus monkey, Chimpanzee, Human
  • Baboon, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3820
  • 3820
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together