Anti-B3GNT5 Rabbit Polyclonal Antibody
ABGEAP9649C
: Abgent
- Antibody type:Primary
- Antigen name:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
- Antigen symbol:B3GNT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Center
- Gene ID:8646
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:TLETKPQRRDQRTVC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CHRD antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 439-467 amino acids from the Central region of human CHRD.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: B3GNT5 (UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse