Anti-PITX1 Rabbit Polyclonal Antibody
ABGEAP7334C
: Abgent
- Antibody type:Primary
- Antigen name:Paired-like homeodomain 1
- Antigen symbol:PITX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Antigen synonyms:Hindlimb-expressed homeobox protein backfoot|Homeobox protein PITX1|BFT|Pituitary homeobox 1|Paired-like homeodomain transcription factor 1|PITX1|PTX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PITX1 (paired-like homeodomain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human