Anti-TXNDC12 Rabbit Polyclonal Antibody
Catalog # ABGEAP4871B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Thioredoxin domain-containing protein 12
- Antigen symbol:TXNDC12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:hAG-1|AGR1|hTLP19|ERP18|TLP19|ERP16|ERP19|PDIA16|AG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TXNDC12 (thioredoxin domain containing 12 (endoplasmic reticulum))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse