Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-GSTT1 Rabbit Polyclonal Antibody
  ABGEAP12899A
 :  Abgent
Anti-GSTT1 Rabbit Polyclonal Antibody
  ABGEAP12899A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Glutathione S-transferase theta 1
  • Antigen symbol:
    GSTT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Antigen synonyms:
    GRB2-Associated Binder 4|Growth Factor Receptor Bound Protein 2-Associated Protein 4|GRB2-Associated Binding Protein Family|Member 4|GRB2-Associated-Binding Protein 2-Like|GAB2-Like|GRB2-Associated Binder 2-Like
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: GSTT1 (glutathione S-transferase theta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human