Anti-CYB5D2 Rabbit Polyclonal Antibody [clone: 163CT48.1.9]
Catalog # ABGEAP11281B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome b5 Domain Containing 2
- Antigen symbol:CYB5D2
- Clonality:Polyclonal
- Clone:163CT48.1.9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Antigen synonyms:eIF-4F 25 kDa subunit|EIF4EL1|EIF4E1|eukaryotic translation initiation factor 4E-like 1|eIF-4E|CBP|eIF4E|EIF4F|mRNA cap-binding protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CYB5D2 (cytochrome b5 domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse