Order Entry
Slovenia
ContactUsLinkComponent
Anti-PIDD Rabbit Polyclonal Antibody
Anti-PIDD Rabbit Polyclonal Antibody
  AVIVARP57269_P050
undefined
Anti-PIDD Rabbit Polyclonal Antibody
  AVIVARP57269_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    p53-induced death domain protein 1
  • Antigen symbol:
    PIDD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    55367
  • Antigen synonyms:
    LRDD|PIDD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: PIDD (p53-induced death domain protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human