Anti-CUX2 Rabbit Polyclonal Antibody
Catalog # AVIVARP47628_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cut-like homeobox 2
- Antigen symbol:CUX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:13048
- Antigen synonyms:Cux-2|Cutl2|1700051K22Rik|ENSMUSG00000072641
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CUX2 (cut-like homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: