Anti-ADAM29 Rabbit Polyclonal Antibody
AVIVARP46991_P050
- Antibody type:Primary
- Antigen name:a disintegrin and metallopeptidase domain 29
- Antigen symbol:ADAM29
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:244486
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ADAM29 (ADAM metallopeptidase domain 29)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: