Anti-HNF4G Rabbit Polyclonal Antibody
Catalog # AVIVARP45616_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:hepatocyte nuclear factor 4, gamma
- Antigen symbol:HNF4G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3174
- Antigen synonyms:NR2A2|NR2A3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HNF4G (hepatocyte nuclear factor 4, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: