Anti-GRHL2 Rabbit Polyclonal Antibody
AVIVARP39635_P050
- Antibody type:Primary
- Antigen name:grainyhead-like transcription factor 2
- Antigen symbol:GRHL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:79977
- Antigen synonyms:BOM|DFNA28|ECTDS|TFCP2L3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GRHL2 (grainyhead-like 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: