Anti-TNFRSF25 Rabbit Polyclonal Antibody
BOSSBS-0421R
: Bioss
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 25
- Antigen symbol:TNFRSF25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Antigen synonyms:DR3|LARD|WSL-LR|TR3|WSL-1|APO-3|TNFRSF12|DDR3|TRAMP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: TNFRSF25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat