Order Entry
Slovenia
ContactUsLinkComponent
HPLC columns, MAbPac™ HIC-20
HPLC columns, MAbPac™ HIC-20
Certificates
undefined
HPLC columns, MAbPac™ HIC-20
Specifications
  • Catalog No:
  • 555-7359
  • 555-7357
  • 555-7358
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Temperature limit:
  • 60 °C
  • 60 °C
  • 60 °C