Total organic carbon (TOC) reagent sets, Test 'N Tube™
Supplier: Hach
Certificates
About this item
Used for determination of Total Organic Carbon (TOC) either by low range, mid range or high range test 'N Tube' method respectively. The set includes digestion vials and reagents for 25 to 50 tests.
- Affordable
- Accurate
- Easy to use
16 mm TNT products can be evaluated on DR900 only.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together