Silica gel 60, irregular 60 - 200 µm for flash chromatography
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:63231-67-4
Formula:SiO₂
MDL Number:MFCD00011232
Boiling Pt:2230 °C
Molecular Weight:60.08 g/mol
Melting Pt:>1600 °C
EINECS:231-545-4
REACH:01-2119379499-16
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Bulk density (tapped):460 - 550 g/l
- pH (5 %):6.5 - 7.5
- Loss on drying (160°C; 15 min):2.0 - 4.0 %
- Particle size ( 63 µm):Max. 5 %
- Particle size ( 200 µm):Max. 5 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC84893.290 spec_VWRC84893.290 spec_VWRC84893.320 spec_VWRC84893.320 spec_VWRC84893.360
Product specification
More
VWR® for chromatography columns, consumables and reagents
Product Literature
Frequently Bought Together