Dissecting scissors, Kilner, Aesculap®
AESCBC168R
: Aesculap
- Type:Curved, flat blunt tips
- Length:150 mm
- Environmentally Preferable:
- Pk:1
Scissors with extra flat tips for dissecting and undermining the skin tissue.
Scissors with extra flat tips for dissecting and undermining the skin tissue.