Order Entry
Slovenia
ContactUsLinkComponent
 
Solid phase extraction cartridges, Sep-Pak® Classic
  WATEWAT051910
 :  WATERS
Solid phase extraction cartridges, Sep-Pak® Classic
  WATEWAT051910
 :  WATERS

 

  • Column Format:
    Cartridge
  • Particle Size:
    55 - 105 µm
  • Capacity:
    0,85 ml
  • Description:
    Sep-Pak® Classic short
  • Seq:
    02
  • Environmentally Preferable:
  • Bed weight:
    360 mg
  • Phase:
    C18
  • Pk:
    50

 

 

Silica-based bonded phase with strong hydrophobicity used to adsorb analytes of even weak hydrophobicity from aqueous solutions.

Typical applications include drugs and their metabolites in serum, plasma or urine, desalting of peptides, trace organics in environmental water samples, organic acids in beverages; similar in behaviour to reversed phase HPLC columns.