Transport swabs, Transwab®
Supplier: MEDICAL WIRE EQUIPMENT
New Product
Certificates
About this item
Transwab with a narrow shaft and bud for nasopharyngeal or urethral sampling.
- Nasopharyngeal sampling such as for Bordetella pertussis
- Open cell flow-through allows full access and release of sampled organisms
- Optimum sample pick-up and release
Used in conventional and automated processing. Compatible with automated systems.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together