Order Entry
Slovenia
ContactUsLinkComponent
Examination gloves, latex, Absogel
Examination gloves, latex, Absogel
Certificates
undefined
Examination gloves, latex, Absogel
Specifications
  • Catalog No:
  • UNIG1470
  • UNIG1480
  • UNIG1475
  • Model:
  • Absogel
  • Absogel
  • Absogel
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Micro-texured
  • Micro-texured
  • Micro-texured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,18 mm
  • 0,18 mm
  • 0,18 mm
  • Finger thickness:
  • 0,28 mm
  • 0,28 mm
  • 0,28 mm
  • Palm thickness:
  • 0,26 mm
  • 0,26 mm
  • 0,26 mm
  • EN norm:
  • EN 455
  • EN 455
  • EN 455