4-Cyano-4'-octyloxybiphenyl ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4'-Octyloxy-[1,1'-biphenyl]-4-carbonitrile
CAS Number:52364-73-5
:
Formula:C₂₁H₂₅NO
MDL Number:MFCD00075145
Molecular Weight:307.44 g/mol
Melting Pt:51…77 °C
Flash Pt:113 °C (235 °F)
EINECS:257-878-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together