Life science sealing films For PCR and storage
: Brand
This film is ideal for PCR, ELISA and sample storage. Due to its adhesive, it can be used down to –80 °C, is DMSO resistant and resistant to common solvents.
- Temperature stability up to 120 °C
- Tight seal for minimising evaporation
- DMSO and solvent resistant
Two of the foils (701376 and 701377) can be removed without leaving any residue so that work can continue unhindered after transport or storage.
Frequently Bought Together