Gas cylinder cabinets, type 90, G-Ultimate
: asecos
Fire resistant type 90 safety cabinets for the safe storage of gas cylinders in buildings in accordance with DIN EN 14470-2.
- Easy installation of equipment and pipes - large interior height (1858 mm)
- Access possibilities on the top of the cabinet
- No unauthorised use - doors lockable with cylinder lock (suitable for locking systems)
- Door opening angle max. 180°
- Simple installation - adjusting aids to compensate for uneven floor
- Quickly put into operation - integrated air ducts ready for connection to a technical exhaust system, complete and uniform ventilation of the cabinet interior from the bottom to the top
The fire protection isolation includes also intumescent joint seal. Sheet steel housing with plastic coating in RAL 7035 (light-grey). Security cabinet for storing gas bottles, fire resistant for up to 90 minutes, it can be installed in outdoor areas. The cabinet is connected to the ventilation system via a standard 75 mm exhaust outlet. The internal height is 1858 mm, enabling the exhaust duct and fittings to be installed easily and conveniently. A wide retractable access ramp enables materials to be handled comfortably and safely.
Conform to laboratory reglementation EN-14727 CE. GS and TüV approved. Made in Germany.
: 90 minutes fire rated
Furnace tested (type tested)
GS approval, CE conformal
Labelling in accordance with EN 14470-2
Furnace tested (type tested)
GS approval, CE conformal
Labelling in accordance with EN 14470-2
: Cabinets are also available in RAL 1004, body warning yellow/doors warning yellow at no extra costs. Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check Avantor.com or contact Avantor for further information.
: Delivery prices available upon request. The gas cylinders and valves are not included in delivery.
Frequently Bought Together