Water quality test strips, 5-in-1
Catalog # HACH2755250
Supplier: Hach
CAS Number:
Specifications
- Pk:50
- Description:5 in 1 water quality test strips
- Environmentally Preferable:
Specifications
About this item
5-in-1 test strips are multi-parameter test strips test offering five water quality measurements on 1 strip.
With Hach water quality test strips, technicians in the field can test many samples in only a few minutes and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical reagent handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening to detect the presence of materials that might interfere with laboratory testing.
Free chlorine (0 to 10 mg/L)
Total chlorine (0 to 10 mg/L)
Total hardness (as CaCO₃, 0 to 425 mg/L and 0 to 428 mg/L)
Total Alkalinity (as CaCO₃, 0 to 240 mg/L)
pH (6,2 to 8,4)