3-Bromobenzaldehyde ethylene acetal ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(3-Bromophenyl)-1,3-dioxolane
CAS Number:17789-14-9
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00003209
Boiling Pt:132…133 °C (8 mmHg)
Density:1.514
Molecular Weight:229.07 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:241-766-8
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together