Geranyl bromide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6138-90-5
:
Formula:C₁₀H₁₇Br
MDL Number:MFCD00000243
Molecular Weight:217.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:228-123-7
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together