Safety cabinets for pressurised gas cylinders, type G30, ECO line plus
Safety cabinets for pressurised gas cylinders-inside storage is made up of sheet steel, powder-coated.
- Classical door technology
- Access control with cylinder lock
- Enhanced ventilation in every cabinet level
- Adjustable feet for level equalisation
: Conforms with DIN EN 14470-2, EK5/AK4 09-10, DIN EN 14727, CE certified, TRGS 510 chapter 10, type G30.
Frequently Bought Together