Size exclusion chromatography column, Superdex™ Peptide 10/300 GL
Catalog # 17-5176-01
Supplier: Cytiva
Specifications
- Flow rate:0,15 - 0,6 ml/min
- Particle Size:13 µm
- Max. pressure:18 bar
- Capacity:24 ml
- Description:Superdex™ Peptide 10/300 GL
- Seq:05
- Environmentally Preferable:
- pH range:1 - 14
- Exclusion limit:100 - 7000 (Mr) globular proteins
- Column hardware:Glass
- Bed dimensions:10×310 mm
- Pk:1
Specifications
About this item
Superdex™ Peptide 10/300 GL is a prepacked column for high resolution size exclusion chromatography for small scale preparative purification and analysis of peptides and other small biomolecules with molecular weights between 100 and 7000.
- High resolution separations of proteins, peptides and other biomolecules according to size
- Well suited for the polishing step in a purification procedure
- Easy and predictable scale-up